Assay Details
Target Gene Details
Entrez Gene ID: | 2650 |
Gene Name: | glucosaminyl (N-acetyl) transferase 1, core 2 |
Gene Aliases: |
C2GNT, C2GNT-L, C2GNT1, G6NT, NACGT2, NAGCT2 |
Location: |
Chr.9:76441666-76507416 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCNT1 | NM_001097634.1 | 1 | 150 | NP_001091103.1 |
| AK313343.1 | ||||
| AY196293.1 | 1 | 150 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520423 | Chr.9:76432201 - 76450383 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map