Assay Details
Target Gene Details
Entrez Gene ID: | 3301 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member A1 |
Gene Aliases: |
DJ-2, DjA1, HDJ2, HSDJ, HSJ-2, HSJ2, HSPF4, NEDD7, hDJ-2 |
Location: |
Chr.9:33025203-33039907 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJA1 | NM_001314039.1 | NP_001300968.1 | ||
| NM_001539.3 | NP_001530.1 | |||
| AJ277242.1 | CAB93148.1 | |||
| AK095541.1 | ||||
| AK289623.1 | ||||
| AK298725.1 | ||||
| AY186741.1 | AAO31694.1 | |||
| BC008182.1 | AAH08182.1 | |||
| BM469197.1 | ||||
| BT007292.1 | AAP35956.1 | |||
| D13388.1 | BAA02656.1 | |||
| L08069.1 | AAC37517.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv614143 | Chr.9:32583509 - 33141754 on Build GRCh38 | Gain |
|
| esv3620303 | Chr.9:32569144 - 33069475 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map