Assay Details
Target Gene Details
Entrez Gene ID: | 105376204 |
Gene Name: | uncharacterized LOC105376204 |
Gene Aliases: |
- |
Location: |
Chr.9:106852964-106863015 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105376204 | XM_017015409.1 | 1 | 1312 | XP_016870898.1 |
Target Gene Details
Entrez Gene ID: | 58499 |
Gene Name: | zinc finger protein 462 |
Gene Aliases: |
Zfp462 |
Location: |
Chr.9:106860154-107012258 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF462 | XM_006717212.3 | XP_006717275.1 |
More Information
Set Membership: |
|

Genomic Map