Assay Details
Target Gene Details
Entrez Gene ID: | 103752585 |
Gene Name: | RORB antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.9:74473435-74498553 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RORB-AS1 | NR_125791.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831622 | Chr.9:74434084 - 74532100 on Build GRCh38 | Gain |
|
| nsv614658 | Chr.9:74454955 - 74539535 on Build GRCh38 | Gain |
|

Genomic Map