Assay Details
Target Gene Details
Entrez Gene ID: | 9719 |
Gene Name: | ADAMTS like 2 |
Gene Aliases: |
GPHYSD1 |
Location: |
Chr.9:133532164-133575519 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMTSL2 | NM_001145320.1 | NP_001138792.1 | ||
| AB011177.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
|
| nsv951799 | Chr.9:133473579 - 133617678 on Build GRCh38 | Deletion |
|
| nsv615637 | Chr.9:133532712 - 133539394 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112155122] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map