Assay Details
Target Gene Details
Entrez Gene ID: | 196528 |
Gene Name: | AT-rich interaction domain 2 |
Gene Aliases: |
BAF200, p200 |
Location: |
Chr.12:45729683-45908037 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARID2 | NM_152641.2 | NP_689854.2 | ||
| XM_006719272.3 | XP_006719335.1 | |||
| AK127872.1 | BAC87171.1 | |||
| AY727870.2 | AAU20329.2 | |||
| CR749833.1 | CAH18689.1 | |||
| DQ096628.1 |
More Information
Set Membership: |
|

Genomic Map