Assay Details
Target Gene Details
Entrez Gene ID: | 3736 |
Gene Name: | potassium voltage-gated channel subfamily A member 1 |
Gene Aliases: |
AEMK, EA1, HBK1, HUK1, KV1.1, MBK1, MK1, RBK1 |
Location: |
Chr.12:4909907-4918256 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNA1 | NM_000217.2 | NP_000208.2 | ||
| DA236475.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483060 | Chr.12:3190835 - 5290834 on Build GRCh38 | Loss |
|
| esv3628373 | Chr.12:4878455 - 4913960 on Build GRCh38 | Gain |
|
| nsv523628 | Chr.12:4907227 - 4913272 on Build GRCh38 | Gain |
|
| nsv1040098 | Chr.12:3987637 - 5646161 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map