Assay Details
Target Gene Details
Entrez Gene ID: | 4938 |
Gene Name: | 2'-5'-oligoadenylate synthetase 1 |
Gene Aliases: |
E18/E16, IFI-4, OIAS, OIASI |
Location: |
Chr.12:112906777-112932190 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OAS1 | NM_001032409.2 | NP_001027581.1 | ||
| NM_001320151.1 | NP_001307080.1 | |||
| NM_002534.3 | NP_002525.2 | |||
| NM_016816.3 | NP_058132.2 | |||
| XM_006719434.1 | XP_006719497.1 | |||
| XM_011538413.1 | XP_011536715.1 | |||
| XM_017019361.1 | XP_016874850.1 | |||
| XM_017019362.1 | XP_016874851.1 | |||
| AJ629455.1 | CAF33358.1 | |||
| AK223006.1 | BAD96726.1 | |||
| AK225384.1 | ||||
| AK291003.1 | ||||
| AK300346.1 | ||||
| AK301498.1 | ||||
| AY730627.1 | AAW63049.1 | |||
| AY730628.1 | AAW63050.1 | |||
| BC000562.2 | ||||
| BC061587.1 | AAH61587.1 | |||
| BC071981.1 | ||||
| BT006785.1 | AAP35431.1 | |||
| D00068.1 | BAA00047.1 | |||
| X02874.1 | CAA26633.1 | |||
| X04371.1 | CAB51602.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832515 | Chr.12:112778024 - 112926931 on Build GRCh38 | Gain |
|
| nsv1042636 | Chr.12:112627854 - 113014974 on Build GRCh38 | Gain |
|
| nsv832516 | Chr.12:112883300 - 113040797 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map