Assay Details
Target Gene Details
Entrez Gene ID: | 144321 |
Gene Name: | GLI pathogenesis-related 1 like 2 |
Gene Aliases: |
- |
Location: |
Chr.12:75391070-75432688 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLIPR1L2 | NM_001270396.1 | NP_001257325.1 | ||
| NM_152436.2 | NP_689649.1 | |||
| NR_072995.1 | ||||
| XM_011537949.2 | XP_011536251.1 | |||
| XM_011537950.2 | XP_011536252.1 | |||
| XM_017018848.1 | XP_016874337.1 | |||
| XM_017018849.1 | XP_016874338.1 | |||
| AK093175.1 | BAC04085.1 | |||
| BC016749.1 | AAH16749.1 | |||
| BC029557.1 | AAH29557.1 | |||
| BX640916.1 | CAE45957.1 |
More Information
Set Membership: |
|

Genomic Map