Assay Details
Target Gene Details
Entrez Gene ID: | 101055625 |
Gene Name: | OVCH1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:29389294-29487324 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OVCH1-AS1 | NR_073170.1 | |||
| NR_073171.1 | ||||
| NR_073172.1 | ||||
| AA910547.1 | ||||
| BQ058205.1 | ||||
| BX119718.1 | ||||
| HY017985.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525100 | Chr.12:29423584 - 29433022 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75619241] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map