Assay Details
Target Gene Details
Entrez Gene ID: | 282809 |
Gene Name: | POC1 centriolar protein B |
Gene Aliases: |
CORD20, PIX1, TUWD12, WDR51B |
Location: |
Chr.12:89419718-89526262 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POC1B | NM_001199777.1 | NP_001186706.1 | ||
| NM_172240.2 | NP_758440.1 | |||
| NR_037659.1 | ||||
| NR_037660.1 | ||||
| AB053266.1 | BAB69057.1 | |||
| AK027829.1 | ||||
| AK074772.1 | BAC11198.1 | |||
| AK123106.1 | ||||
| AK123625.1 | BAC85666.1 | |||
| AK295596.1 | ||||
| AL832918.1 | CAH10775.1 | |||
| BC026080.2 | AAH26080.1 | |||
| BX648293.1 | 1 | 1502 | ||
| EF011619.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580371 | Chr.12:89421998 - 89431934 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs148325985] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map