Assay Details
Target Gene Details
Entrez Gene ID: | 56341 |
Gene Name: | protein arginine methyltransferase 8 |
Gene Aliases: |
HRMT1L3, HRMT1L4 |
Location: |
Chr.12:3381349-3593973 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRMT8 | NM_001256536.1 | NP_001243465.1 | ||
| AK315619.1 | ||||
| KR014345.1 |
Target Gene Details
Entrez Gene ID: | 100129223 |
Gene Name: | thyroid cancer-associated transcript 155 |
Gene Aliases: |
- |
Location: |
Chr.12:3483712-3492819 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THCAT155 | NR_126055.1 | 2 | 4646 | |
| AK125333.1 | 2 | 4646 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483060 | Chr.12:3190835 - 5290834 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map