Assay Details
Target Gene Details
Entrez Gene ID: | 101928002 |
Gene Name: | uncharacterized LOC101928002 |
Gene Aliases: |
- |
Location: |
Chr.12:69713633-69738568 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928002 | NR_110072.1 |

Genomic Map