Assay Details
Target Gene Details
Entrez Gene ID: | 8099 |
Gene Name: | cyclin dependent kinase 2 associated protein 1 |
Gene Aliases: |
DOC1, DORC1, ST19, doc-1, p12DOC-1 |
Location: |
Chr.12:123260970-123272316 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK2AP1 | NM_001270433.1 | NP_001257362.1 | ||
| NM_004642.3 | NP_004633.1 | |||
| NR_073007.1 | ||||
| NR_073008.1 | ||||
| AB006077.1 | BAA22937.1 | |||
| AF006484.1 | AAC77831.1 | |||
| BC034717.1 | AAH34717.1 | |||
| BP290637.1 | ||||
| CB964037.1 | ||||
| DR759851.1 |
More Information
Set Membership: |
|

Genomic Map