Assay Details
Target Gene Details
Entrez Gene ID: | 11103 |
Gene Name: | KRR1, small subunit processome component homolog |
Gene Aliases: |
HRB2, RIP-1 |
Location: |
Chr.12:75497639-75511638 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRR1 | NM_007043.6 | NP_008974.5 | ||
| AK313687.1 | ||||
| BC005225.1 | ||||
| BC016778.1 | AAH16778.1 | |||
| BC026107.2 | AAH26107.1 | |||
| BC033887.2 | AAH33887.1 | |||
| EF010919.1 | ||||
| U55766.1 | AAB00557.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470305 | Chr.12:75502908 - 75593580 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map