Assay Details
Target Gene Details
Entrez Gene ID: | 3875 |
Gene Name: | keratin 18 |
Gene Aliases: |
CK-18, CYK18, K18 |
Location: |
Chr.12:52948871-52952901 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT18 | NM_000224.2 | NP_000215.1 | ||
| NM_199187.1 | NP_954657.1 | |||
| AK129587.1 | ||||
| AK223093.1 | BAD96813.1 | |||
| AK308506.1 | ||||
| AK313988.1 | ||||
| AY762101.1 | AAX07828.1 | |||
| BC000180.2 | AAH00180.1 | |||
| BC000698.2 | AAH00698.1 | |||
| BC004253.1 | AAH04253.1 | |||
| BC008636.1 | AAH08636.1 | |||
| BC020982.1 | AAH20982.1 | |||
| BC072017.1 | AAH72017.1 | |||
| BG753529.1 | ||||
| BT019412.1 | AAV38219.1 | |||
| CD106591.1 | ||||
| X12881.1 | CAA31375.1 | |||
| X12883.1 | CAA31377.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832416 | Chr.12:52779656 - 52971582 on Build GRCh38 | Loss |
|
| nsv973068 | Chr.12:52950527 - 52952866 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map