Assay Details
Target Gene Details
Entrez Gene ID: | 84790 |
Gene Name: | tubulin alpha 1c |
Gene Aliases: |
TUBA6, bcm948 |
Location: |
Chr.12:49227926-49273338 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBA1C | NM_001303114.1 | NP_001290043.1 | ||
| NM_001303115.1 | NP_001290044.1 | |||
| NM_001303116.1 | NP_001290045.1 | |||
| NM_001303117.1 | NP_001290046.1 | |||
| NM_032704.4 | NP_116093.1 | |||
| AK223024.1 | BAD96744.1 | |||
| AK293589.1 | ||||
| AL539200.3 | ||||
| BC004949.1 | AAH04949.1 | |||
| BC005946.1 | AAH05946.1 | |||
| BC011790.1 | AAH11790.1 | |||
| BC019298.2 | AAH19298.1 | |||
| BC021088.1 | AAH21088.1 | |||
| BC033064.1 | AAH33064.1 | |||
| BC051297.1 | AAH51297.1 | |||
| BC063036.1 | AAH63036.1 | |||
| BG469587.1 | ||||
| DA686435.1 | ||||
| DC366449.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv976611 | Chr.12:49268633 - 49270486 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map