Assay Details
Target Gene Details
Entrez Gene ID: | 4938 |
Gene Name: | 2'-5'-oligoadenylate synthetase 1 |
Gene Aliases: |
E18/E16, IFI-4, OIAS, OIASI |
Location: |
Chr.12:112906777-112932190 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OAS1 | NM_001320151.1 | NP_001307080.1 | ||
| AA085948.1 | ||||
| AJ629455.1 | CAF33358.1 | |||
| DQ823040.1 | ||||
| DQ914956.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560260 | Chr.12:112925887 - 112928886 on Build GRCh38 | Gain+Loss |
|
| nsv560259 | Chr.12:112924602 - 112927776 on Build GRCh38 | Gain |
|
| esv25722 | Chr.12:112922509 - 112929506 on Build GRCh38 | Gain |
|
| nsv560258 | Chr.12:112922497 - 112928244 on Build GRCh38 | Loss |
|
| nsv560263 | Chr.12:112926577 - 112929077 on Build GRCh38 | Loss |
|
| nsv1047747 | Chr.12:112917435 - 113003116 on Build GRCh38 | Gain |
|
| nsv1042636 | Chr.12:112627854 - 113014974 on Build GRCh38 | Gain |
|
| nsv832516 | Chr.12:112883300 - 113040797 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map