Assay Details
Target Gene Details
Entrez Gene ID: | 144715 |
Gene Name: | RAD9 checkpoint clamp component B |
Gene Aliases: |
- |
Location: |
Chr.12:110501655-110534208 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
More Information
Set Membership: |
|

Genomic Map