Assay Details
Target Gene Details
Entrez Gene ID: | 3747 |
Gene Name: | potassium voltage-gated channel subfamily C member 2 |
Gene Aliases: |
KV3.2 |
Location: |
Chr.12:75040078-75209815 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNC2 | NM_001260497.1 | NP_001247426.1 | ||
| NM_001260498.1 | NP_001247427.1 | |||
| NM_001260499.1 | NP_001247428.1 | |||
| NM_139136.3 | NP_631874.1 | |||
| NM_139137.3 | NP_631875.1 | |||
| NM_153748.2 | NP_715624.1 | |||
| XM_005268855.3 | XP_005268912.1 | |||
| XM_006719382.3 | XP_006719445.1 | |||
| XM_006719383.3 | XP_006719446.1 | |||
| XM_006719384.3 | XP_006719447.1 | |||
| XM_006719385.3 | XP_006719448.1 | |||
| XM_017019282.1 | XP_016874771.1 | |||
| AB209994.1 | ||||
| AF268896.1 | AAL27272.1 | |||
| AF268897.2 | AAL27273.1 | |||
| AK094720.1 | BAC04407.1 | |||
| AK294269.1 | ||||
| AK309245.1 | ||||
| AY118169.1 | AAM81577.1 | |||
| AY243473.1 | AAO89503.1 | |||
| BC093635.1 | AAH93635.1 | |||
| BC111991.1 | AAI11992.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559420 | Chr.12:75025938 - 75085599 on Build GRCh38 | Loss |
|
| nsv522290 | Chr.12:74941247 - 75120843 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map