Assay Details
Target Gene Details
Entrez Gene ID: | 100885789 |
Gene Name: | IFNG antisense RNA 1 |
Gene Aliases: |
GS1-410F4.2, NEST, Tmevpg1 |
Location: |
Chr.12:67989445-68021327 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFNG-AS1 | NR_104124.1 | |||
| NR_104125.1 | ||||
| AK124066.1 | ||||
| BG756823.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039638 | Chr.12:67899571 - 68006091 on Build GRCh38 | Gain |
|

Genomic Map