Assay Details
Target Gene Details
Entrez Gene ID: | 56899 |
Gene Name: | ankyrin repeat and sterile alpha motif domain containing 1B |
Gene Aliases: |
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2 |
Location: |
Chr.12:98729904-99984727 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Target Gene Details
Entrez Gene ID: | 101928937 |
Gene Name: | uncharacterized LOC101928937 |
Gene Aliases: |
- |
Location: |
Chr.12:99093359-99105011 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928937 | NR_110095.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv847 | Chr.12:99065597 - 99100208 on Build GRCh38 | Insertion |
|
| dgv297e214 | Chr.12:99051318 - 99128190 on Build GRCh38 | Gain |
|
| nsv559933 | Chr.12:99042804 - 99399467 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map