Assay Details
Target Gene Details
Entrez Gene ID: | 3034 |
Gene Name: | histidine ammonia-lyase |
Gene Aliases: |
HIS, HSTD |
Location: |
Chr.12:95972662-95996365 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAL | NM_001258333.1 | NP_001245262.1 | ||
| NM_001258334.1 | NP_001245263.1 | |||
| NM_002108.3 | NP_002099.1 | |||
| XM_011538249.2 | XP_011536551.1 | |||
| XM_017019246.1 | XP_016874735.1 | |||
| AK297004.1 | ||||
| AK298736.1 | ||||
| AK303544.1 | ||||
| BC096096.1 | AAH96096.1 | |||
| BC096097.3 | AAH96097.1 | |||
| BC096098.3 | AAH96098.1 | |||
| BC096099.1 | AAH96099.1 | |||
| D16626.1 | BAA04047.1 |
More Information
Set Membership: |
|

Genomic Map