Assay Details
Target Gene Details
Entrez Gene ID: | 59277 |
Gene Name: | netrin 4 |
Gene Aliases: |
PRO3091 |
Location: |
Chr.12:95657807-95790758 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTN4 | NM_021229.3 | NP_067052.2 | ||
| AF119916.3 | AAF69670.2 | |||
| AF278532.1 | AAG30822.1 | |||
| AF297711.1 | AAG53651.1 | |||
| AK024691.1 | BAB14964.1 | |||
| AK226070.1 | ||||
| AK290591.1 | ||||
| AK315796.1 | ||||
| AL137540.1 | CAB70800.3 | |||
| AL833767.1 | CAH56243.1 | |||
| AY330211.1 | AAP92113.1 | |||
| BC013591.1 | AAH13591.2 | |||
| BC136798.1 | ||||
| BC136799.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833 | Chr.12:95665257 - 95696754 on Build GRCh38 | Insertion |
|
| nsv1052350 | Chr.12:95528912 - 95686094 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map