Assay Details
Target Gene Details
Entrez Gene ID: | 5908 |
Gene Name: | RAP1B, member of RAS oncogene family |
Gene Aliases: |
K-REV, RAL1B |
Location: |
Chr.12:68610839-68660605 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAP1B | NM_001010942.2 | NP_001010942.1 | ||
| NM_001251917.1 | NP_001238846.1 | |||
| NM_001251918.1 | NP_001238847.1 | |||
| NM_001251921.1 | NP_001238850.1 | |||
| NM_001251922.1 | NP_001238851.1 | |||
| NM_015646.5 | NP_056461.1 | |||
| AB062128.1 | BAB93460.1 | |||
| AF279894.1 | AAK07551.1 | |||
| AJ420497.1 | ||||
| AK298818.1 | ||||
| AK301401.1 | ||||
| AK301428.1 | ||||
| AK311559.1 | ||||
| AK312371.1 | ||||
| AK316447.1 | ||||
| AK316543.1 | ||||
| AL080212.1 | CAB45777.1 | |||
| BC000176.3 | AAH00176.1 | |||
| BC078173.1 | AAH78173.1 | |||
| BC095467.1 | AAH95467.1 | |||
| BG717530.1 | ||||
| DB448914.1 | ||||
| X08004.1 | CAB46488.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952200 | Chr.12:68612321 - 68643920 on Build GRCh38 | Duplication |
|
| nsv522941 | Chr.12:68497196 - 68787063 on Build GRCh38 | Gain |
|
| esv3629852 | Chr.12:68611952 - 68623274 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76217285] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map