Assay Details
Target Gene Details
Entrez Gene ID: | 102216268 |
Gene Name: | chondrogenesis-associated transcript |
Gene Aliases: |
CISTR-ACT, CISTRACT, re52431 |
Location: |
Chr.12:53750447-53757034 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CISTR | NR_104332.1 | |||
| HY042141.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1116700 | Chr.12:53753803 - 53753907 on Build GRCh38 | Tandem Duplication |
|
| esv3302703 | Chr.12:53753805 - 53753920 on Build GRCh38 | Tandem Duplication |
|

Genomic Map