Assay Details
Target Gene Details
Entrez Gene ID: | 283349 |
Gene Name: | Ras association domain family member 3 |
Gene Aliases: |
RASSF5 |
Location: |
Chr.12:64610513-64697567 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASSF3 | NM_178169.3 | NP_835463.1 | ||
| NR_040718.1 | ||||
| XM_011538195.2 | XP_011536497.1 | |||
| AY217662.1 | AAO61687.1 | |||
| AY217663.1 | AAO61688.1 | |||
| BC100949.2 | AAI00950.1 | |||
| BC100950.1 | AAI00951.1 | |||
| BC100951.1 | AAI00952.1 | |||
| BC100952.1 | AAI00953.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832443 | Chr.12:64485076 - 64663485 on Build GRCh38 | Loss |
|
| nsv832444 | Chr.12:64587704 - 64774692 on Build GRCh38 | Loss |
|
| dgv2682n54 | Chr.12:64609189 - 64806142 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77945625] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map