Assay Details
Target Gene Details
Entrez Gene ID: | 55206 |
Gene Name: | strawberry notch homolog 1 |
Gene Aliases: |
MOP3, Sno |
Location: |
Chr.12:123289109-123365204 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SBNO1 | NM_001167856.1 | NP_001161328.1 | ||
| NM_018183.3 | NP_060653.3 | |||
| XM_005253572.3 | XP_005253629.1 | |||
| XM_005253573.4 | XP_005253630.1 | |||
| XM_005253575.3 | XP_005253632.1 | |||
| XM_005253576.3 | XP_005253633.1 | |||
| XM_006719473.3 | XP_006719536.1 | |||
| XM_006719474.3 | XP_006719537.1 | |||
| XM_011538533.2 | XP_011536835.1 | |||
| XM_011538534.2 | XP_011536836.1 | |||
| XM_011538535.2 | XP_011536837.1 | |||
| XM_017019556.1 | XP_016875045.1 | |||
| XM_017019557.1 | XP_016875046.1 | |||
| XM_017019558.1 | XP_016875047.1 | |||
| XM_017019559.1 | XP_016875048.1 | |||
| AB014772.1 | BAB19784.1 | |||
| AK001563.1 | BAA91758.1 | |||
| AK074256.1 | ||||
| AK096864.1 | ||||
| BC133704.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs138323775] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map