Assay Details
Target Gene Details
Entrez Gene ID: | 10376 |
Gene Name: | tubulin alpha 1b |
Gene Aliases: |
K-ALPHA-1 |
Location: |
Chr.12:49127782-49131521 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBA1B | NM_006082.2 | NP_006073.2 | ||
| AK026516.1 | ||||
| AK026632.1 | ||||
| AK056693.1 | 1 | 294 | ||
| AK057773.1 | ||||
| AK094717.1 | 1 | 959 | ||
| AK098357.1 | ||||
| AK098635.1 | ||||
| AK098771.1 | ||||
| AK289354.1 | ||||
| BC000696.2 | AAH00696.1 | |||
| BC001128.1 | AAH01128.1 | |||
| BC006379.2 | AAH06379.1 | |||
| BC006481.1 | AAH06481.1 | |||
| BC008659.2 | AAH08659.1 | |||
| BC009314.2 | AAH09314.1 | |||
| BC009509.2 | AAH09509.1 | |||
| BC009512.2 | AAH09512.1 | |||
| BC009513.2 | AAH09513.1 | |||
| BC010494.1 | AAH10494.1 | |||
| BC011572.1 | AAH11572.1 | |||
| BC015883.2 | AAH15883.1 | |||
| BC017004.1 | AAH17004.1 | |||
| BC021564.1 | AAH21564.1 | |||
| BC030820.1 | AAH30820.1 | |||
| BC071904.1 | AAH71904.1 | |||
| DQ400107.1 | ABD60581.1 | |||
| K00558.1 | AAA91576.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv975485 | Chr.12:49127627 - 49131900 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map