Assay Details
Target Gene Details
Entrez Gene ID: | 55681 |
Gene Name: | SCY1 like pseudokinase 2 |
Gene Aliases: |
CVAK104 |
Location: |
Chr.12:100267137-100341724 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCYL2 | NM_001317784.1 | NP_001304713.1 | ||
| NM_017988.5 | NP_060458.3 | |||
| XM_017019618.1 | XP_016875107.1 | |||
| XM_017019619.1 | XP_016875108.1 | |||
| XM_017019620.1 | XP_016875109.1 | |||
| AK000936.1 | BAA91433.1 | |||
| AK125091.1 | ||||
| AK292980.1 | ||||
| BC063798.1 | AAH63798.1 | |||
| DA234206.1 | ||||
| DB127407.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559964 | Chr.12:99872025 - 100538490 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs144836561] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map