Assay Details
Target Gene Details
Entrez Gene ID: | 784 |
Gene Name: | calcium voltage-gated channel auxiliary subunit beta 3 |
Gene Aliases: |
CAB3, CACNLB3 |
Location: |
Chr.12:48814432-48828943 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNB3 | NM_001206917.1 | NP_001193846.1 | ||
| AK122911.1 | 1 | 518 | ||
| AK300901.1 | ||||
| AK304537.1 | ||||
| AL702978.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv698 | Chr.12:48795148 - 48828796 on Build GRCh38 | Insertion |
|
| nsv826363 | Chr.12:48783444 - 48857206 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map