Assay Details
Target Gene Details
Entrez Gene ID: | 57613 |
Gene Name: | family with sequence similarity 234 member B |
Gene Aliases: |
KIAA1467 |
Location: |
Chr.12:12927680-13083449 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM234B | XM_017019706.1 | XP_016875195.1 |
Target Gene Details
Entrez Gene ID: | 55507 |
Gene Name: | G protein-coupled receptor class C group 5 member D |
Gene Aliases: |
- |
Location: |
Chr.12:12940518-12955908 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPRC5D | XM_005253421.3 | XP_005253478.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745567 | Chr.12:12246086 - 13092998 on Build GRCh38 | Deletion |
|
| nsv832333 | Chr.12:12896803 - 13108130 on Build GRCh38 | Gain |
|
| nsv520173 | Chr.12:12930203 - 12972003 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs148632130] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map