Assay Details
Target Gene Details
Entrez Gene ID: | 6579 |
Gene Name: | solute carrier organic anion transporter family member 1A2 |
Gene Aliases: |
OATP, OATP-A, OATP1A2, SLC21A3 |
Location: |
Chr.12:21264600-21403657 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLCO1A2 | NM_021094.3 | 14 | 7065 | NP_066580.1 |
| NM_134431.3 | 16 | 7535 | NP_602307.1 | |
| XM_005253474.3 | 15 | 6956 | XP_005253531.1 | |
| XM_011520818.1 | 15 | 6963 | XP_011519120.1 | |
| XM_011520819.1 | 15 | 7110 | XP_011519121.1 | |
| BX093727.1 | 1 | 161 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv515735 | Chr.12:20857114 - 21267778 on Build GRCh38 | Gain+Loss |
|
| esv3628799 | Chr.12:20916259 - 21333930 on Build GRCh38 | Loss |
|
| dgv1404n100 | Chr.12:20996197 - 21267778 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map