Assay Details
Target Gene Details
Entrez Gene ID: | 8315 |
Gene Name: | BRCA1 associated protein |
Gene Aliases: |
BRAP2, IMP, RNF52 |
Location: |
Chr.12:111642146-111686023 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRAP | NM_006768.4 | NP_006759.3 | ||
| XM_005253944.4 | XP_005254001.1 | |||
| XM_011538789.2 | XP_011537091.1 | |||
| XM_017019992.1 | XP_016875481.1 | |||
| XM_017019993.1 | XP_016875482.1 | |||
| XM_017019994.1 | XP_016875483.1 | |||
| XM_017019995.1 | XP_016875484.1 | |||
| AB208878.1 | BAD92115.1 | |||
| AF035620.1 | AAC24200.1 | |||
| AF035950.1 | AAB88538.1 | |||
| AK290379.1 | ||||
| AK299326.1 | ||||
| AK308279.1 | ||||
| AY332222.1 | AAP93638.1 | |||
| BC136698.1 | ||||
| BC136699.1 |
More Information
Set Membership: |
|

Genomic Map