Assay Details
Target Gene Details
Entrez Gene ID: | 51474 |
Gene Name: | LIM domain and actin binding 1 |
Gene Aliases: |
EPLIN, SREBP3 |
Location: |
Chr.12:50175780-50283570 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIMA1 | NM_001113546.1 | NP_001107018.1 | ||
| NM_001113547.1 | NP_001107019.1 | |||
| NM_016357.4 | NP_057441.1 | |||
| XM_011538455.2 | XP_011536757.1 | |||
| XM_017019430.1 | XP_016874919.1 | |||
| AB209512.1 | BAD92749.1 | |||
| AF198454.1 | AAF23755.1 | |||
| AF198455.1 | AAF23756.1 | |||
| AK000335.1 | BAA91092.1 | |||
| AK000372.1 | BAA91120.1 | |||
| AK222971.1 | BAD96691.1 | |||
| AK297766.1 | ||||
| AK314447.1 | ||||
| AL136911.1 | CAB66845.1 | |||
| BC001247.1 | AAH01247.2 | |||
| BC110815.1 | AAI10816.1 | |||
| BC136763.1 | ||||
| DA738195.1 |
More Information
Set Membership: |
|

Genomic Map