Assay Details
Target Gene Details
Entrez Gene ID: | 51350 |
Gene Name: | keratin 76 |
Gene Aliases: |
HUMCYT2A, KRT2B, KRT2P |
Location: |
Chr.12:52768155-52777345 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT76 | NM_015848.4 | NP_056932.2 | ||
| AK299390.1 | ||||
| M99063.1 | AAA35746.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv976622 | Chr.12:52776354 - 52777554 on Build GRCh38 | Duplication |
|
| esv3629524 | Chr.12:52688070 - 52835680 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map