Assay Details
Target Gene Details
Entrez Gene ID: | 2004 | 
Gene Name: | ELK3, ETS transcription factor | 
Gene Aliases: | 
	
		ERP, NET, SAP-2, SAP2 | 
Location: | 
		
			Chr.12:96194382-96269835 on Build GRCh38 | 
		Assay Gene Location: | Within Intron 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| ELK3 | NM_005230.3 | NP_005221.2 | ||
| XM_006719275.3 | XP_006719338.1 | |||
| AK312694.1 | ||||
| BC017371.1 | AAH17371.1 | |||
| DA754301.1 | ||||
| Z36715.1 | CAA85309.1 | 
More Information
Set Membership: | 
		   
					
						
							 | 
				

Genomic Map