Assay Details
Target Gene Details
Entrez Gene ID: | 51367 |
Gene Name: | POP5 homolog, ribonuclease P/MRP subunit |
Gene Aliases: |
HSPC004, RPP2, RPP20, hPop5 |
Location: |
Chr.12:120579045-120581452 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POP5 | NM_015918.3 | NP_057002.2 | ||
| NM_198202.1 | NP_937845.1 | |||
| XM_011538441.1 | XP_011536743.1 | |||
| AF070660.1 | AAD20966.1 | |||
| AF117232.1 | AAF17213.1 | |||
| AJ306296.1 | CAC59822.1 | |||
| AK223206.1 | BAD96926.1 | |||
| AK303144.1 | ||||
| BC012505.1 | AAH12505.1 | |||
| BI668578.1 | ||||
| CB995722.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv907 | Chr.12:120580294 - 120614874 on Build GRCh38 | Insertion |
|
| nsv906 | Chr.12:120535880 - 120580638 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs146429057] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map