Assay Details
Target Gene Details
Entrez Gene ID: | 84660 |
Gene Name: | coiled-coil domain containing 62 |
Gene Aliases: |
CT109, ERAP75, TSP-NY |
Location: |
Chr.12:122773621-122827385 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC62 | NM_201435.4 | NP_958843.2 | ||
| NR_027918.2 | ||||
| XM_006719643.1 | XP_006719706.1 | |||
| XM_006719644.1 | XP_006719707.1 | |||
| XM_011538856.1 | XP_011537158.1 | |||
| XM_011538857.1 | XP_011537159.1 | |||
| XM_017020033.1 | XP_016875522.1 | |||
| AK097663.1 | ||||
| AK124633.1 | BAC85909.1 | |||
| AK292466.1 | ||||
| AY254201.1 | AAP13075.1 | |||
| BC060796.1 | AAH60796.1 | |||
| DB458565.1 |
More Information
Set Membership: |
|

Genomic Map