Assay Details
Target Gene Details
Entrez Gene ID: | 84660 |
Gene Name: | coiled-coil domain containing 62 |
Gene Aliases: |
CT109, ERAP75, TSP-NY |
Location: |
Chr.12:122773621-122827385 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC62 | XM_017020033.1 | 1 | 359 | XP_016875522.1 |
More Information
Set Membership: |
|

Genomic Map