Assay Details
Target Gene Details
Entrez Gene ID: | 4257 |
Gene Name: | microsomal glutathione S-transferase 1 |
Gene Aliases: |
GST12, MGST, MGST-I |
Location: |
Chr.12:16347142-16377189 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGST1 | NM_001267598.1 | NP_001254527.1 | ||
| XM_011520674.2 | XP_011518976.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1049805 | Chr.12:16284461 - 16498558 on Build GRCh38 | Loss |
|
| esv3892150 | Chr.12:16339581 - 16384735 on Build GRCh38 | Loss |
|
| nsv832338 | Chr.12:16319300 - 16487163 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map