Assay Details
Target Gene Details
Entrez Gene ID: | 3845 |
Gene Name: | KRAS proto-oncogene, GTPase |
Gene Aliases: |
C-K-RAS, CFC2, K-RAS2A, K-RAS2B, K-RAS4A, K-RAS4B, KI-RAS, KRAS1, KRAS2, NS, NS3, RALD, RASK2 |
Location: |
Chr.12:25204789-25252093 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRAS | NM_004985.4 | NP_004976.2 | ||
| NM_033360.3 | NP_203524.1 | |||
| XM_006719069.3 | XP_006719132.1 | |||
| XM_011520653.2 | XP_011518955.1 | |||
| XM_017019292.1 | XP_016874781.1 | |||
| XM_017019293.1 | XP_016874782.1 | |||
| AF493917.1 | AAM12631.1 | |||
| AK292510.1 | ||||
| BC013572.2 | AAH13572.1 | |||
| BT007153.1 | AAP35817.1 | |||
| M35504.1 | AAA35689.1 | |||
| M35505.1 | AAA35690.1 | |||
| M54968.1 | AAB41942.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557798 | Chr.12:25206907 - 25244348 on Build GRCh38 | Loss |
|
| nsv470279 | Chr.12:25231724 - 25246187 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map