Assay Details
Target Gene Details
Entrez Gene ID: | 57097 |
Gene Name: | poly(ADP-ribose) polymerase family member 11 |
Gene Aliases: |
ARTD11, C12orf6, MIB006 |
Location: |
Chr.12:3808861-3873448 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARP11 | NM_001286521.1 | NP_001273450.1 | ||
| NM_001286522.1 | NP_001273451.1 | |||
| NM_020367.5 | NP_065100.2 | |||
| NR_104461.1 | ||||
| XM_005253711.4 | XP_005253768.1 | |||
| XM_011520970.1 | XP_011519272.1 | |||
| XM_011520971.2 | XP_011519273.1 | |||
| XM_011520973.2 | XP_011519275.1 | |||
| XM_017019670.1 | XP_016875159.1 | |||
| XM_017019671.1 | XP_016875160.1 | |||
| AF263540.1 | AAF91391.1 | |||
| AK299372.1 | ||||
| AK314916.1 | ||||
| BC017569.1 | AAH17569.1 | |||
| BX640960.1 | CAE45979.1 | |||
| BX647487.1 | ||||
| CR749294.1 | CAH18149.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv468981 | Chr.12:3576597 - 3989342 on Build GRCh38 | Gain |
|
| nsv483060 | Chr.12:3190835 - 5290834 on Build GRCh38 | Loss |
|
| nsv832313 | Chr.12:3689562 - 3906773 on Build GRCh38 | Loss |
|
| nsv1036828 | Chr.12:3607219 - 3846326 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map