Assay Details
Target Gene Details
Entrez Gene ID: | 1840 |
Gene Name: | deltex E3 ubiquitin ligase 1 |
Gene Aliases: |
hDx-1 |
Location: |
Chr.12:113056711-113098028 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DTX1 | NM_004416.2 | NP_004407.2 | ||
| XM_011538009.2 | XP_011536311.1 | |||
| AF053700.1 | AAC06246.1 | |||
| BC048216.1 | AAH48216.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519540 | Chr.12:113042705 - 113459599 on Build GRCh38 | Gain+Loss |
|
| nsv560264 | Chr.12:113050137 - 113136469 on Build GRCh38 | Loss |
|
| esv3630792 | Chr.12:112988703 - 113838810 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map