Assay Details
Target Gene Details
Entrez Gene ID: | 55206 |
Gene Name: | strawberry notch homolog 1 |
Gene Aliases: |
MOP3, Sno |
Location: |
Chr.12:123289109-123365204 on Build GRCh38 |
Assay Gene Location: | Within Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SBNO1 | NM_001167856.1 | 31 | 8814 | NP_001161328.1 |
| NM_018183.3 | 31 | 8811 | NP_060653.3 | |
| XM_005253572.3 | 32 | 8952 | XP_005253629.1 | |
| XM_011538534.2 | 31 | 8850 | XP_011536836.1 | |
| XM_011538535.2 | 31 | 8852 | XP_011536837.1 | |
| XM_017019556.1 | 32 | 8936 | XP_016875045.1 | |
| XM_017019557.1 | 32 | 8943 | XP_016875046.1 | |
| XM_017019558.1 | 33 | 9006 | XP_016875047.1 | |
| XM_017019559.1 | 32 | 9073 | XP_016875048.1 |
More Information
Set Membership: |
|

Genomic Map