Assay Details
Target Gene Details
Entrez Gene ID: | 6334 |
Gene Name: | sodium voltage-gated channel alpha subunit 8 |
Gene Aliases: |
CERIII, CIAT, EIEE13, MED, NaCh6, Nav1.6, PN4 |
Location: |
Chr.12:51589959-51812865 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCN8A | XM_006719556.3 | XP_006719619.1 | ||
| AY510083.1 | AAS89692.1 |
More Information
Set Membership: |
|

Genomic Map