Assay Details
Target Gene Details
Entrez Gene ID: | 3856 |
Gene Name: | keratin 8 |
Gene Aliases: |
CARD2, CK-8, CK8, CYK8, K2C8, K8, KO |
Location: |
Chr.12:52897187-52949866 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT8 | NM_001256282.1 | NP_001243211.1 | ||
| NM_001256293.1 | NP_001243222.1 | |||
| NM_002273.3 | NP_002264.1 | |||
| NR_045962.1 | ||||
| AK222941.1 | BAD96661.1 | |||
| AK290938.1 | ||||
| AK310257.1 | ||||
| AK315826.1 | ||||
| BC000654.2 | ||||
| BC008200.1 | AAH08200.1 | |||
| BC011373.1 | AAH11373.2 | |||
| BC063513.1 | ||||
| BC073760.1 | AAH73760.1 | |||
| BC075839.1 | AAH75839.1 | |||
| CX166336.1 | ||||
| M34225.1 | AAA35748.1 | |||
| M77025.1 | ||||
| U76549.1 | AAB18966.1 | |||
| X12882.1 | CAA31376.1 | |||
| X74929.1 | CAA52882.1 | |||
| X98614.1 | CAA67203.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv558965 | Chr.12:52862019 - 52913242 on Build GRCh38 | Loss |
|
| nsv832416 | Chr.12:52779656 - 52971582 on Build GRCh38 | Loss |
|
| nsv952181 | Chr.12:52896117 - 52903416 on Build GRCh38 | Deletion |
|
| esv3581638 | Chr.12:52883196 - 52908855 on Build GRCh38 | Gain |
|
| nsv983328 | Chr.12:52897420 - 52902354 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map