Assay Details
Target Gene Details
| Entrez Gene ID: | 2004 | 
| Gene Name: | ELK3, ETS transcription factor | 
| Gene Aliases: | ERP, NET, SAP-2, SAP2 | 
| Location: | Chr.12:96194382-96269835 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| ELK3 | NM_005230.3 | NP_005221.2 | ||
| XM_006719275.3 | XP_006719338.1 | |||
| AK312694.1 | ||||
| BC017371.1 | AAH17371.1 | |||
| DA754301.1 | ||||
| Z36715.1 | CAA85309.1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  non-DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top