Assay Details
Target Gene Details
Entrez Gene ID: | 100874249 |
Gene Name: | DENND5B antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:31589923-31615351 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DENND5B-AS1 | NR_046909.1 | |||
| DA250246.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760267 | Chr.12:31473798 - 31756974 on Build GRCh38 | Gain+Loss |
|
| nsv832365 | Chr.12:31489877 - 31655598 on Build GRCh38 | Loss |
|

Genomic Map