Assay Details
Target Gene Details
Entrez Gene ID: | 613227 |
Gene Name: | HIG1 hypoxia inducible domain family member 1C |
Gene Aliases: |
GM921 |
Location: |
Chr.12:50936223-50972592 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIGD1C | XM_011538649.2 | XP_011536951.1 | ||
| XM_017019783.1 | XP_016875272.1 | |||
| XM_017019785.1 | XP_016875274.1 |
Target Gene Details
Entrez Gene ID: | 4891 |
Gene Name: | solute carrier family 11 member 2 |
Gene Aliases: |
AHMIO1, DCT1, DMT1, NRAMP2 |
Location: |
Chr.12:50952263-51028335 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC11A2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3629495 | Chr.12:50946886 - 50973141 on Build GRCh38 | Loss |
|
| esv34039 | Chr.12:50932919 - 51278127 on Build GRCh38 | Loss |
|
| nsv1053466 | Chr.12:50956502 - 50982186 on Build GRCh38 | Loss |
|

Genomic Map